MilliporeSigma
GC Analysis of Volatiles in Roasted Coffee Beans on Omegawax after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber
GC Analysis of Volatiles in Roasted Coffee Beans on Omegawax after SPME using a 50/30 μm DVB/Carboxen/PDMS Fiber application for SPME, suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Omegawax Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS)
needle size 24 ga, StableFlex, for use with autosampler
mobile phase component
Product No.
Description
Pricing
Ethyl acetate
anhydrous, 99.8%
related product
Product No.
Description
Pricing
SPME Fiber Holder
for use with manual sampling
SPME Fiber Holder
for use with CTC CombiPAL and Gerstel MPS2
standard
Product No.
Description
Pricing
p-Cymene
analytical standard
Methyl acetate
analytical standard
α-Terpinene
≥95.0% (GC)
γ-Terpinene
analytical standard
2,3-Butanedione
analytical standard
Pyridine
anhydrous, 99.8%
2,5-Dimethylpyrrole
98%
3,4-Hexanedione
95%
2,3-Dihydro-5-methylfuran
97%
2,5-Dimethylfuran
99%
3-Ethylpyridine
≥98%, FG
2-Ethylpyrazine
≥98%, FG
3-Penten-2-one
≥70%
2-Methylpyrazine
≥99%
Acetoin
primarily dimer, ≥95%, FG
Ethyl alcohol, Pure
200 proof, anhydrous, ≥99.5%
2-Acetyl-1-methylpyrrole
≥98%, FG
Pyrrole-2-carboxaldehyde
98%
2-Butanone solution
certified reference material, 2000 μg/mL in methanol: water (9:1)
Linalool
analytical standard
Linalyl acetate
analytical standard
2,3-Hexanedione
analytical standard
2,3-Pentanedione
analytical standard
Pyrazine
analytical standard
Furfural
analytical standard
Hexanal
analytical standard
5-Methylfurfural
analytical standard
Furfuryl acetate
analytical reference material
Pyrrole
analytical standard
Furfuryl alcohol
analytical standard
2-Methylfuran
analytical standard
2,5-Dimethylpyrazine
analytical standard
γ-Butyrolactone
analytical standard
CONDITIONS
sample/matrix
2 g of coffee beans
SPME fiber
50/30 μm divinylbenzene/Carboxen on polydimethylsiloxane on a StableFlex fiber (57329-U)
extraction
headspace, with a pre-equilibration period of 10 min. at 60 °C, and then a fiber exposure of 40 min. at 60 °C, agitated in an alternate clockwise-anticlockwise rotation mode at 500 rpm
desorption process
260 °C for 5 min
column
Omegawax 250, 30 m x 0.25 mm I.D., 0.25 μm (24136)
oven
40 °C (5 min), 4 °C/min to 230 °C, 50 °C/min to 280 °C (2 min)
inj. temp.
260 °C
detector
MS, m/z 40-400
carrier gas
helium, 34.8 cm/sec constant
injection
splitless
scan range
m/z 40-400
sample
Roasted coffee
liner
0.75 mm I.D. SPME/Splitless (2633901)
Description
Analysis Note
Headspace-SPME followed by GC analysis was used in this application to analyze volatile compounds in roasted coffee. SPME is ideal for analyzing the headspace of solid samples like coffee beans. The Carboxen/DVB/PDMS fiber is the fiber of choice for analyzing the wide range of analytes commonly found in roast coffee beans.The relatively polar Omegawax 250 GC column provided high resolution.
Legal Information
null