MilliporeSigma
US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.00 μm
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.00 μm
SLB®-5ms Capillary GC Column
L × I.D. 15 m × 0.10 mm, df 0.10 μm
SLB®-5ms Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 0.18 μm
accessory
Product No.
Description
Pricing
Wheaton wide-mouth bottle
capacity 32 oz (1000 mL), amber, white polypropylene cap, PTFE flat liner
Wheaton straight-mouth bottles with cap, clear
capacity 8 oz (250 mL), white polypropylene cap, PTFE flat liner
guard column
Product No.
Description
Pricing
Intermediate Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.25 mm
Intermediate Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.53 mm
recommended
Product No.
Description
Pricing
Polyimide Sealing Resin
pkg of 5 g
required but not provided
Product No.
Description
Pricing
"Y" GlasSeal Connector, Borosilicate Glass
pkg of 1 ea
"Y" GlasSeal Connector, Fused Silica
pkg of 1 ea
"Y" GlasSeal Connector, Fused Silica
pkg of 3 ea
standard
Product No.
Description
Pricing
Underground Storage Tank (UST) Modified DRO
certified reference material, TraceCERT®, 1000 μg/mL each component in hexane, ampule of 1 mL
CONDITIONS
column
SLB-5ms, 20 m x 0.18 mm I.D., 0.18 μm (28564-U)
oven
50 °C (2 min), 20 °C/min to 325 °C (1 min)
inj. temp.
250 °C
detector
FID, 350 °C
carrier gas
helium, 0.9 mL/min, constant
injection
1 μL, splitless (splitter open at 0.5 min)
liner
2 mm I.D., FocusLiner inlet liner
sample
each analyte at 50 μg/mL in methylene chloride
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany