GC Analysis of Verbenone Enantiomers on Astec® CHIRALDEX™ B-TA
![GC Analysis of Verbenone Enantiomers on Astec® CHIRALDEX™ B-TA suitable for GC](/deepweb/assets/sigmaaldrich/product/images/233/932/df88ec4e-dedf-4f17-a52f-d0e12c4d4edd/800/df88ec4e-dedf-4f17-a52f-d0e12c4d4edd.jpg)
CONDITIONS
column
Astec CHIRALDEX B-TA, 30 m x 0.25 mm I.D., 0.12 μm (73023AST)
oven
120 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
1 μL, 80:1 split
sample
2.5 mg/mL
peak 1: S(-)-verbenone
peak 2: R(+)-verbenone
Description
Analysis Note
Can reverse elution order by using CHIRALDEX G-TA.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC