MilliporeSigma
GC Analysis of Trichloroanisole (TCA) and Precursors in Red Wine on SLB®-5ms after SPME using 100 μm PDMS Fiber
GC Analysis of Trichloroanisole (TCA) and Precursors in Red Wine on SLB®-5ms after SPME using 100 μm PDMS Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Polydimethylsiloxane (PDMS)
metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
CONDITIONS
sample/matrix
1.5 mL red wine + 600 μL 5% potassium carbonate + 240 mg sodium chloride + 60 μL acetic anhydride
SPME fiber
metal fiber coated with 100 μm PDMS (57928-U)
extraction
headspace, 50 °C for 30 min., with stirring
desorption process
250 °C for 3 min
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
50 °C (1 min), 25 °C/min to 280 °C
detector
ECD, 290 °C
carrier gas
helium, 1.5 mL/min, constant
liner
0.75 mm I.D. SPME
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany