MilliporeSigma
GC Analysis of TCEP Test Mix on SLB®-IL100, 110 °C Oven Temp.
GC Analysis of TCEP Test Mix on SLB®-IL100, 110 °C Oven Temp. suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-IL100 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SLB-IL100, 30 m x 0.25 mm I.D., 0.20 μm (28884-U)
oven
110 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec
injection
1.0 μL 100:1 split
liner
4 mm I.D., split, cup design
sample
(TCEP Test Mix (N08682), each analyte at various concentrations in isooctane)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany