MilliporeSigma
GC Analysis of Phthalate Esters in Corn Oil (Spiked) on SLB®-5ms after SPME with a 100 μm PDMS Fiber
GC Analysis of Phthalate Esters in Corn Oil (Spiked) on SLB®-5ms after SPME with a 100 μm PDMS Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 0.18 μm
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly
7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
SPME Fiber Assembly
100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
SPME Fiber Assembly
100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, plain red hub
related product
Product No.
Description
Pricing
Vials, screw top, clear glass (vial only)
volume 15 mL, clear glass vial, thread for 18-400, pkg of 100 ea
Screw cap, phenolic, with open center
black phenolic hole cap, for use with 15 mL vial with 18-400 thread, pkg of 100 ea
Septa, tan PTFE/silicone
white tan PTFE/silicone, diam. × thickness 16 mm × 0.090 in., for use with 15 mL vial, pkg of 100 ea
SPME Fiber Holder
for use with Manual Sampling, pk of 1
SPME Fiber Holder
for use with CTC CombiPAL, Gerstel MPS2 Autosamplers, pk of 1
standard
Product No.
Description
Pricing
Diphenyl phthalate
PESTANAL®, analytical standard
Dicyclohexyl phthalate
PESTANAL®, analytical standard
Bis(methylglycol) phthalate
analytical standard
Dimethyl phthalate solution
certified reference material, 5000 μg/mL in methanol
Bis(2-ethylhexyl) phthalate
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Dibutyl phthalate
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Diisobutyl phthalate
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
CONDITIONS
sample/matrix
corn oil spiked with phthalate esters at 500 μg/kg
SPME fiber
100 μm PDMS (57300-U)
extraction
headspace, 95 °C (measured in headspace)
desorption process
260 °C for 4 min
column
F.S. CAP. SLB-5ms 20 m x 0.18 mm I.D., 0.18 μm (28564-U)
oven
60 °C (1 min), 10 °C/min to 330 °C (10 min)
inj. temp.
260 °C
carrier gas
helium, 0.6 mL/min, constant
liner
0.75 mm I.D. SPME
sample
Corn oil
Description
Analysis Note
In this application, SPME was used for the analysis of phthalate esters from oily samples, specifically the oil-based flavor packets included with ramen noodle kits. Spiked samples of corn oil and flavored oils included with ramen noodle kits were analyzed using high temperature SPME headspace extraction using a 100 μm PDMS fiber which was then directly desorbed into the GC/MS system. Calibration standards were prepared in corn oil, and spiked samples of chicken and beef oil-based flavors were analyzed. The SLB-5ms GC columns provide consistently low-bleed, inert, efficient, and durable separations.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany