MilliporeSigma
GC Analysis of Fuel Oil #6 on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Fuel Oil #6 on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-1 Capillary GC Column
L × I.D. 15 m × 0.10 mm, df 0.10 μm
CONDITIONS
column
Equity-1, 15 m x 0.10 mm I.D., 0.10 μm (28039-U)
oven
80 °C, 50 °C/min to 325 °C
inj. temp.
250 °C
detector
FID, 350 °C
carrier gas
hydrogen, 45 cm/sec constant
injection
0.3 μL, 100:1 split, 0.1 min pre-injection dwell time
liner
2 mm I.D., straight
sample
No.6 Fuel Oil standard, 20 mg/mL in hexane:chloroform (47536-U)
Description
Legal Information
null