MilliporeSigma
GC Analysis of Fluoxetine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX B-DA
GC Analysis of Fluoxetine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX™ B-DA suitable for GC
Materials
standard
Product No.
Description
Pricing
Fluoxetine hydrochloride
solid
S-(+)-Fluoxetine hydrochloride
≥98% (HPLC), solid
R-(−)-Fluoxetine hydrochloride
>98% (HPLC), solid
Fluoxetine hydrochloride
VETRANAL®, analytical standard
Fluoxetine-d5 hydrochloride
98 atom % D
CONDITIONS
column
Astec CHIRALDEX B-DA, 10 m x 0.25 mm I.D., 0.12 μm (72021AST)
oven
175 °C
detector
FID, 250 °C
carrier gas
helium, 15 psi
inj. temp.
250 °C
sample
peaks 1 & 2: fluoxetine enantiomers (N-TFA derivatives)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC