GC Analysis of FAMEs in Cod Liver Oil on SLB®-IL100
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL100, 30 m x 0.25 mm I.D., 0.20 μm (28884-U)
oven
120 °C, 3.0 °C/min to 240 °C
inj. temp.
240 °C
detector
FID, 240 °C
carrier gas
hydrogen, 35 cm/sec constant
injection
1 μL 50:1 split
sample
cod liver oil FAMEs
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany