MilliporeSigma
GC Analysis of FAMEs in B20 Biodiesel (Soy Source) on Omegawax
GC Analysis of FAMEs in B20 Biodiesel (Soy Source) on Omegawax suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Omegawax Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
standard
Product No.
Description
Pricing
Omegawax Column Test Mix
50 μg/mL in hexane, analytical standard
CONDITIONS
column
Omegawax 250, 30 m x 0.25 mm I.D., 0.25 μm (24136)
oven
50 °C, 13 °C/min to 270 °C (5 min)
inj. temp.
250 °C
detector
FID, 270 °C
detector
helium, 40 cm/sec
injection
1 μL 100:1 split
liner
4 mm I.D. FocusLiner inlet liner (no taper)
sample
B20 biodiesel (soy source) diluted 1:20 in n-hexane
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
null