MilliporeSigma
GC Analysis of cis C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SLB®-IL111 after HPLC Fractionation using Silver Ion
GC Analysis of cis C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SLB®-IL111 after HPLC Fractionation using Silver Ion suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-IL111 Capillary GC Column
L × I.D. 100 m × 0.25 mm, df 0.20 μm
related product
Product No.
Description
Pricing
Inlet Liner, Split Type, Cup Design (unpacked)
pkg of 1 ea
CONDITIONS
column
SLB-IL111, 100 m x 0.25 mm I.D., 0.20 μm (29647-U)
oven
168 °C isothermal
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
hydrogen, 1 mL/min
injection
1 μL, 100:1 split
liner
4 mm I.D., split liner with cup (2051001)
sample
cis fraction of partially hydrogenated vegetable oil FAMEs, prepared using Ag-ion fractionation
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany