MilliporeSigma
GC Analysis of β-Butyrolactone Enantiomers on Astec® CHIRALDEX B-DM
GC Analysis of β-Butyrolactone Enantiomers on Astec® CHIRALDEX™ B-DM suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-DM Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
β-Butyrolactone
98%
CONDITIONS
column
Astec CHIRALDEX B-DM, 30 m x 0.25 mm I.D., 0.12 μm (77023AST)
oven
70 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peaks 1 & 2: β-butyrolactone
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC