MilliporeSigma
GC Analysis of a 37-Component FAME Mix on Omegawax
GC Analysis of a 37-Component FAME Mix on Omegawax suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Omegawax Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
standard
Product No.
Description
Pricing
F.A.M.E. Mix, C4-C24
certified reference material,  wt. % (varied)
Supelco 37 Component FAME Mix
certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL
CONDITIONS
column
Omegawax 250, 30 m x 0.25 mm I.D., 0.25 μm (24136)
oven
50 °C, 3.0 °C/min to 280 °C
inj. temp.
240 °C
detector
FID, 280 °C
carrier gas
helium, 40 cm/sec constant
injection
1 μL 50:1 split
sample
Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
Description
Legal Information
null