MilliporeSigma
GC Analysis of a 10-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of a 10-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 10 m × 0.10 mm, df 0.10 μm
CONDITIONS
column
SLB-5ms, 10 m x 0.10 mm I.D., 0.10 μm (28465-U)
oven
40 °C (1 min), 80 °C/min to 160 °C, 5 °C/min to 340 °C (2 min)
inj. temp.
280 °C
MSD interface
280 °C
scan range
45-470 m/z
carrier gas
helium, 0.52 mL/min
injection
0.5 μL 10:1 split
sample
10-component pesticide mix, each analyte at 0.8-2.0 μg/mL in hexane:acetone (50:50)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany