MilliporeSigma
GC Analysis of 2-Butanol Enantiomers (Underivatized) on Astec® CHIRALDEX G-TA
GC Analysis of 2-Butanol Enantiomers (Underivatized) on Astec® CHIRALDEX™ G-TA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-TA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
2-Butanol
analytical standard
(S)-(+)-2-Butanol
99%
(R)-(−)-2-Butanol
99%
2-Butanol
anhydrous, 99.5%
2-Butanol
≥99%
2-Butanol
JIS special grade, ≥99.0%
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
gradient
30 °C (5 min), 5 °C/min to 110 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
1 μL, 80:1 split
sample
2 mg/mL each racemate in methylene chloride
peak 1: S(+)-2-butanol
peak 2: R(-)-2-butanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC