MilliporeSigma
US EPA Method 624: GC Analysis of Volatiles on VOCOL® (60 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap
US EPA Method 624: GC Analysis of Volatiles on VOCOL® (60 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap suitable for GC
Materials
analytical column
Product No.
Description
Pricing
VOCOL® Capillary GC Column
L × I.D. 60 m × 0.53 mm, df 3.00 μm
CONDITIONS
sample/matrix
20 ppb each component in 5 mL water (25381)
purge trap
VOCARB 3000 (21066-U)
purge
35 mL/min for 11 min
dry purge
3 min
desorption temp.
250 °C for 4 min
bake
260 °C for 10 min
column
VOCOL®
, 60 m × 0.53 mm I.D.,3.0 μm (25381)
oven
35 °C (4 min), 6 °C/min to 200 °C
scan range
m/z 35-260 at 0.6 sec/scan
carrier gas
helium, 7.5 mL/min
Description
Legal Information
VOCOL is a registered trademark of Merck KGaA, Darmstadt, Germany