MilliporeSigma
GC Analysis of FAMEs on SP®-2330
GC Analysis of FAMEs on SP®-2330 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SP®-2330 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SP-2330, 30 m × 0.25 mm I.D., 0.20 μm (24019)
oven
200 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 20 cm/sec @ 200 °C
injection
0.5 μL, split 100:1
sample
FAMEs from a natural source dissolved in chloroform
Description
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC