MilliporeSigma
GC Analysis of cis/trans FAME Column Performance Mix on SP®-2560 per AOAC Method 2012.13
GC Analysis of cis/trans FAME Column Performance Mix on SP®-2560 per AOAC Method 2012.13 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SP®-2560 Capillary GC Column
L × I.D. 100 m × 0.25 mm, df 0.20 μm
standard
Product No.
Description
Pricing
cis/trans F.A.M.E Column Performance Mix
2.5 mg/mL in dichloromethane, ampule of 1 mL
CONDITIONS
column
SP-2560, 100 m x 0.25 mm I.D., 0.20 μm (24056)
oven
60 °C (1 min), 15 °C/min to 165 °C (1 min), 2 °C/min to 225 °C (20 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 0.8 mL/min
injection
1 μL, 10:1 split
liner
4 mm I.D., split/splitless type, wool packed single taper FocusLiner design
sample
cis/trans FAME Column Performance Mix (40495-U)
Description
Analysis Note
Prior to analysis of samples, AOAC Method 2012.13 requires that system resolution be verified to ensure sufficient separation of the C18:1 cis/trans isomers. This is done by injecting a cis/trans FAME column performance mix (40495-U), a qualitative mix that contains a mixture of C18:1 trans and C18:1 cis isomers. Resolution is calculated between two groups of FAME isomers. Group 1 = C18:1Δ13t and C18:1Δ14t, whereas group 2 = C18:1Δ9c (oleic acid) and C18:1Δ10c. A resolution value of >1.0 is considered acceptable. As shown, the resolution value calculated using the SP-2560 column is R=1.12.
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SP is a registered trademark of Sigma-Aldrich Co. LLC