MilliporeSigma
GC Analysis of C2-C7 Free Fatty Acids on Nukol
GC Analysis of C2-C7 Free Fatty Acids on Nukol suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Nukol Capillary GC Column
size × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
Nukol, 30 m × 0.25 mm I.D., 0.25 μm (24107)
oven
185 °C
inj. temp.
220 °C
detector
FID, 250 °C
carrier gas
helium, 20 cm/sec @ 185 °C
injection
1 μL, 100:1 split
sample
Volatile Acid Standard Mix (46975-U), each analyte at 10 mM in deionized water
Description
Legal Information
null