MilliporeSigma
Dynein light chain 1 phosphorylation controls macropinocytosis.
The Journal of biological chemistry (2004-10-27)
Zhibo Yang, Ratna K Vadlamudi, Rakesh Kumar
ABSTRACT
Recent studies have identified dynein light chain-1 (DLC1), a component of the dynein motor, as a p21-activated kinase 1 (Pak1)-interacting substrate with binding sites mapped to amino acids 61-89 of DLC1 and phosphorylation site at serine 88. Here we investigated the role of DLC1 phosphorylation by Pak1 upon the process of macropinocytosis. We found that Pak1 associates with dynein motor and that Pak1-DLC1 interaction starts at the initiation of pinosome formation and persists in early and late endosomes. Pak1 phosphorylation of DLC1 on Ser-88 controls vesicle formation and trafficking functions, as Ser-88 substitution for alanine prevents macropinocytosis. A peptide spanning the C-terminal 19-amino acid region of DLC1 efficiently blocked Ser-88 phosphorylation and macropinocytosis. These results suggest that the regulation of DLC1 by Pak1 is a novel mechanism by which a signaling kinase might influence macropinocytosis.