MilliporeSigma
  • Field comparison of passive samplers versus UV-photometric analyser to measure surface ozone in a Mediterranean area.
Field comparison of passive samplers versus UV-photometric analyser to measure surface ozone in a Mediterranean area.
Journal of environmental monitoring : JEM (2007-06-08)
J M Delgado Saborit, V J Esteve Cano
ABSTRACT
The aim of the present work is to compare the performance of the Radiello passive sampler versus UV-photometric ozone analyser to measure surface ozone in a Mediterranean Spanish coastal area. The comparison presented considers precision, bias, accuracy, selectivity, detection limit, cost and applicability. For assessing precision, co-located samplers were exposed in duplicate in two reference-sampling sites, beside UV-photometric ozone analyser. Bias was calculated comparing results of passive samplers exposed in three reference-sampling sites and two contrast-sampling sites with the measurements given by the reference analysers. Accuracy was calculated following the EN 482:1994 standard. The limit of detection was calculated as 3 times the standard deviation of the blanks in a batch of passive samplers. The compared Radiello passive samplers give a precision of 5.2%, a bias of 13.8%, an accuracy of 20.5% and a limit of detection of 12.6 microg m(-3). The selectivity and applicability of this methodology is in both aspects successful. Surface ozone levels measured with passive samplers were comparable with the averaged values measured with the reference analyser both in the reference-sampling sites and in the contrast-sampling sites.
Product Description
radiello Diffusive Bodies, white, configured for general use, pk of 20
radiello On-Field Thermometer, Reader, pk of 1
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello BTEX/VOC Starter Kit, CS2 Desorption
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
radiello Hydrogen Sulfide Starter Kit
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Filtration Kit, pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Triangular Support Plate, pk of 20
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Barcode Labels, pk of 198
radiello Anesthetic Gases and Vapor Sampler
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Outdoor Shelter, pk of 10
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20