MilliporeSigma
  • Monitoring of volatile organic compounds in non-residential indoor environments.
Monitoring of volatile organic compounds in non-residential indoor environments.
Indoor air (2008-04-24)
P Bruno, M Caselli, G de Gennaro, S Iacobellis, M Tutino
ABSTRACT
A weekly monitoring campaign of volatile organic compounds (VOC), with single sampling of 24 h, was carried out in non-residential indoor environments such as libraries, pharmacies, offices, gymnasiums, etc., in order to evaluate the VOC concentrations to which people are exposed. Moreover, an outdoor sample was coupled to each indoor site to point out the influence of indoor sources. They were sampled with Radiello diffusive samplers for thermal desorption and analyzed by GC-MS. As already described in other papers, the VOC levels of most of the indoor sites were higher than that observed in the corresponding outdoor sites. For example, some sites showed a level of pollution that is ten times higher than their corresponding outdoor site. The monitored environments that had higher concentrations of the investigated VOC were the pharmacies, a newspaper stand, a copy center, and the coffee shops. Analysis of the weekly average concentrations of each pollutant and the use of literature allowed pointing out some site-specific characteristics that singled out possible sources of VOC. These results were verified analyzing the indoor-outdoor ratio (I/O) too. Newspaper stands were characterized by very high concentrations of toluene and pharmacies were characterized by high concentrations of aromatic compounds. Indoor air pollution caused by volatile organic compounds (VOC) might affect human health at home as well as in public and commercial buildings. The main VOC sources in indoor environments are human activities, personal care products, smoking, house cleaning products, building products, and outside pollution. To preserve human health it is necessary to evaluate the average concentrations of VOC to which people are exposed and to identify the main sources of indoor pollution by means of suitable indoor monitoring campaigns in several environments. These investigations allow pointing out the characteristic critical situations of some indoor environments or some other types of environments.
Product Description
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello Diffusive Bodies, white, configured for general use, pk of 20
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello On-Field Thermometer, Reader, pk of 1
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Filtration Kit, pk of 20
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
radiello Hydrogen Sulfide Starter Kit
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Anesthetic Gases and Vapor Sampler
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Outdoor Shelter, pk of 10
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Triangular Support Plate, pk of 20
radiello Barcode Labels, pk of 198
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20