MilliporeSigma
  • Modified 3-alkyl-1,8-dibenzylxanthines as GTP-competitive inhibitors of phosphoenolpyruvate carboxykinase.
Modified 3-alkyl-1,8-dibenzylxanthines as GTP-competitive inhibitors of phosphoenolpyruvate carboxykinase.
Bioorganic & medicinal chemistry letters (2003-09-25)
Louise H Foley, Ping Wang, Pete Dunten, Gwendolyn Ramsey, Mary-Lou Gubler, Stanley J Wertheimer
ABSTRACT
The first non-substrate like inhibitors of human cytosolic phosphoenolpyruvate carboxykinase (PEPCK) competitive with GTP are reported. An effort to discover orally active compounds that improve glucose homeostasis in Type 2 diabetics by reversibly inhibiting PEPCK led to the discovery of 1-allyl-3-butyl-8-methylxanthine (5). We now report modifications at N-1 and C-8 that improved the in vitro activity of the initial xanthine HTS hit by 100-fold and a developing SAR for this class of inhibitor.
Product Description
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm