MilliporeSigma
  • Non-genomic effects of progestins--inhibition of cell growth and increased intracellular levels of cyclic nucleotides.
Non-genomic effects of progestins--inhibition of cell growth and increased intracellular levels of cyclic nucleotides.
The Journal of steroid biochemistry and molecular biology (2003-03-22)
Georg Sager, Anne Ørbo, Ragnhild Jaeger, Catharina Engström
ABSTRACT
The anti-proliferative effect of progestins was studied in human transformed cell lines from the uterine cervix (C-4I, C33A and Me-180). Progestins caused a concentration-dependent inhibition of proliferation. The maximum tested concentration (2.6-3.2 microM) inhibited C-4I cell growth by the following order of potency: progesterone (56%) > medroxyprogesterone (38%) > megestrol acetate (25%). The sensitivity, expressed as I(25) (the concentration that caused 25% inhibition of growth), showed the same order: progesterone (7.7 nM) > medroxyprogesterone (78 nM) > megestrol acetate (570 nM). The intracellular levels of cGMP and cAMP were elevated and the cellular export of these cyclic nucleotides was inhibited by a similar order of potency. The C-4I cell line was devoid of progesterone-, estrogen-, androgen- and glucocorticoid-receptors. In addition, the antiprogestins mifepristone, onapristone and ZK-112993 did not block the anti-proliferative effect of progesterone. On the other hand, antiprogestins (2.3 nM) appeared to have some progesterone-like ("mimetic") activity with inhibition of C-4I cell growth; mifepristone (11%), onapristone (12%) and ZK-112993 (16%). The observed effects of progestins and antiprogestins on C-4I cells were also presented in C33A cells (16% androgen receptor positive) and Me-180 cells (22% progesterone receptor positive, 9% androgen receptor positive and 17% glucocorticoid receptor positive). This study suggests that a non-genomic mechanism contributes to the anti-proliferative effect of progestins.
Product Description
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea