MilliporeSigma
  • Plasticity of nodose ganglion neurons after capsaicin- and vagotomy-induced nerve damage in adult rats.
Plasticity of nodose ganglion neurons after capsaicin- and vagotomy-induced nerve damage in adult rats.
Neuroscience (2010-03-04)
V Ryu, Z Gallaher, K Czaja
ABSTRACT
Previous reports show that vagal afferent innervation of the stomach eventually regenerates from surviving nodose ganglion (NG) neurons after subdiaphragmatic vagotomy. Systemic capsaicin treatment destroys gastric vagal afferent neurons expressing vanilloid receptor 1 (VR1). However, it is not known whether gastric innervation lost after neuronal destruction can be restored. Here, we report that capsaicin-induced damage of NG neurons innervating the stomach in adult rats is followed by restoration of vagal afferent projections. Specifically, we compared measures of neuronal plasticity in NG and vagi after subdiaphragmatic vagotomy or capsaicin treatment. The numbers of VR1-immunoreactive neurons projecting to the stomach were significantly reduced 10 days after either capsaicin treatment or vagotomy. However, the VR1-immunoreactive afferent innervation of the stomach was restored to levels exceeding those of vagotomized rats by 37 days after capsaicin, whereas neither total afferent innervation nor VR1-immunoreactive innervation reached control levels, even by 67 days after vagotomy. Capsaicin treatment significantly increased NG neuronal nitric oxide synthase (nNOS) immunoreactivity at 10 days after capsaicin, and this increase was sustained for the duration of the study, indicating higher nNOS demand in restoration of vagal projections. Vagotomy was associated with a much smaller increase in the number of nNOS-immunoreactive NG neurons, detectable only at 10 days after surgery. The number of nNOS-immunopositive gastric-projecting neurons was dramatically reduced 10 days after either capsaicin treatment or vagotomy but returned to the control level in both groups at 67 days. We found a significantly higher number of growth cones in capsaicin-treated animals compared with controls. Capsaicin significantly increased the number of nNOS-immunopositive and nNOS-immunonegative growth cones in NG at all time points. Vagotomy did not increase the number of nNOS(-) growth cones in NG. We conclude that capsaicin treatment may result in more significant restorative capacities than vagotomy, mainly because of sprouting of capsaicin-insensitive nerve fibers.
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
ECO TWEEN® 80, viscous liquid
Sigma-Aldrich
TWEEN® 80, viscous liquid