MilliporeSigma
  • Prospective multicenter evaluation of the morphometric D-score for prediction of the outcome of endometrial hyperplasias.
Prospective multicenter evaluation of the morphometric D-score for prediction of the outcome of endometrial hyperplasias.
The American journal of surgical pathology (2001-06-23)
J P Baak, A Ørbo, P J van Diest, M Jiwa, P de Bruin, M Broeckaert, W Snijders, P J Boodt, G Fons, C Burger, R H Verheijen, P W Houben, H S The, P Kenemans
ABSTRACT
Prospective multicenter evaluation of the WHO classification and the morphometric D-score to predict endometrial hyperplasia cancer progression. In 132 endometrial hyperplasias WHO classification was performed by two experienced gynecologic pathologists. The D-score was assessed blindly by technicians in a routine diagnostic setting. Development of endometrial carcinoma during a 1-10-year follow-up was used as the end point. Eleven of 132 patients (8%), 10 of 61 (16%) atypical hyperplasias, and 1 of 71 (1%) nonatypical hyperplasias developed cancer. Twenty-six curettings had a D-score < or = 0 ("unfavorable" or endometrial intraepithelial neoplasia) of which 10 (38%) developed cancer. None of the 86 cases with a D-score > 1 ("favorable") and one of the 20 (5%) cases with 0 < D-score < or = 1 ("uncertain") developed cancer. Sensitivity of the D-score was 100%, specificity 82%, the positive and negative predictive values were 38% and 100%, respectively. These values are similar to those in three prior retrospective D-score studies but higher than the WHO values (which are 91%, 58%, 16%, and 99%, respectively). The D-score in endometrial hyperplasias is a more sensitive and specific marker for cancer prediction than the WHO classification, can be assessed in a routine clinical setting on standard hematoxylin and eosin sections (15-30 minutes per case), and is highly reproducible and cost-effective (U.S. $50 per case).
Product Description
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea