MilliporeSigma
  • Rapid and simple method for direct determination of several amphetamines in seized tablets by GC--FID.
Rapid and simple method for direct determination of several amphetamines in seized tablets by GC--FID.
Forensic science international (2005-06-28)
Blagoj Mitrevski, Zoran Zdravkovski
ABSTRACT
A simple and rapid method for direct simultaneous determination of amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxymethamphetamine (MDMA), 3,4-methylenedioxy-N-ethylamphetamine (MDEA) and N-methyl-1-(3,4-methylenedioxyphenyl)-2-butanamine (MBDB) in seized tablets was developed using gas chromatography with flame ionization detection. Separation of all six underivatized amphetamines, including diphenylamine as internal standard, was performed in about 6 min, using SPB-50 capillary column. Amphetamine and methamphetamine eluted with negligible tailing while the other amphetamines had highly symmetrical peaks. Sensitivity per component on-column was in the nanogram range, and reproducibility from 2.6 to 6.6% at low concentration (2.4 microg/mL) and from 1.2 to 2.6% at high (70 microg/mL) concentration. The method has a wide linear range, from Limit of detection (LOD) to almost 200 microg/mL, thus allowing analysis of different samples across a wide range of possible concentrations of amphetamines. This simple, fast and precise method using gas chromatography--flame ionization detector (GC--FID), in conjunction with other methods (TLC, IR, HPLC), can be used for identification of amphetamines and direct determination in seized tablets, especially in laboratories with heavy workload.
Product Description
Supelco
SPB®-50 Capillary GC Column, df 0.25 μm, L × I.D. 30 m × 0.25 mm
Supelco
SPB®-50 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm