MilliporeSigma
  • Facile synthesis of hierarchical porous carbon from crude biomass for high-performance solid-phase microextraction.
Facile synthesis of hierarchical porous carbon from crude biomass for high-performance solid-phase microextraction.
Journal of chromatography. A (2018-03-21)
Hu Cheng, Yang Song, Fang Wang, Rongting Ji, Xiaona Li, Yuhao Fu, Xinglun Yang, Yongrong Bian, Xin Jiang
ABSTRACT
Porous materials have great prospective applications for solid-phase microextraction (SPME) technology because of their large specific surface area and pore volume. In this study, a hierarchical porous carbon (HPC) was synthesized by simple hydrothermal reaction and potassium hydroxide (KOH) activation of crude biomass and found to be an efficient adsorbent for SPME of organic pollutants. Results show that the as-prepared HPC has a partly graphitic amorphous-like structure with ultrahigh specific surface area (2551 m2/g), high pore volume (1.53 cm3/g), good pore size distribution (PSD) (mesopore/micropore ratio of 68%), and great thermal stability (>450 °C). When we utilized it as SPME fiber coating, the extraction capacities for chlorobenzenes (CBs), polychlorinated biphenyls (PCBs), polycyclic aromatic hydrocarbons (PAHs), and phthalates (PAEs) were, respectively, 1.13-39.46, 2.40-7.78, 1.34-36.02, and 1.50-1.83 times higher than those of a commercial polydimethylsiloxane (PDMS) fiber. Under the optimized extraction conditions, an analytical method for CBs with low detection limits (0.01-0.24 ng/L), good repeatability (1.00%-4.93% for intra-day, 1.11%-6.94% for inter-day), and great reproducibility (1.48%-8.91%, n = 3) was developed. Moreover, we evaluated the practicality of the developed method for environmental water sample and obtained satisfactory recoveries (86.21%-104.34%). The findings provide a novel and promising HPC from crude biomass using a low-cost and facile synthetic route for SPME applications.
Product Description
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
2,3,3′-Trichlorobiphenyl, BCR®, certified reference material
Sigma-Aldrich
N-Acetyl-D-penicillamine, for HPLC derivatization, ≥99.0% (T)
Supelco
SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm(Coating thickness includes 65 μm coating + 10 μm OC (overcoating), needle size 23 ga, pkg of 3 ea, fiber L 1 cm
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler