MilliporeSigma
  • Measurement of benzene concentration in urban air using passive sampling.
Measurement of benzene concentration in urban air using passive sampling.
Analytical and bioanalytical chemistry (2011-12-14)
Sylwia Król, Bożena Zabiegała, Jacek Namieśnik
ABSTRACT
The concentration of benzene in urban air in the Tri-City area of Poland (Gdańsk-Sopot-Gdynia, and Tczew) was assessed using diffusive passive samplers (Radiello). Samples were collected during a four-year monitoring campaign (2007-2010) at selected monitoring stations managed by the Agency of Regional Air Quality Monitoring in the Gdańsk Metropolitan Area (ARMAAG) Foundation. The performance of the passive samplers was investigated in a field study that measured the benzene concentration in urban air. The results obtained by the Radiello samplers were compared with the results obtained using an on-line monitor (Chrompack CP 7001). Statistical analysis of the results obtained by the two different techniques (passive and on-line) was performed by a linear regression method (Student's t-test). The influence of temperature fluctuations on the uptake rate behavior of the passive samplers was also investigated.
Product Description
radiello Diffusive Bodies, white, configured for general use, pk of 20
radiello On-Field Thermometer, Reader, pk of 1
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello BTEX/VOC Starter Kit, CS2 Desorption
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
radiello Hydrogen Sulfide Starter Kit
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Filtration Kit, pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Triangular Support Plate, pk of 20
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Barcode Labels, pk of 198
radiello Anesthetic Gases and Vapor Sampler
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Outdoor Shelter, pk of 10
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20