MilliporeSigma
  • Development and testing of species-specific ELISA assays to measure IFN-γ and TNF-α in bottlenose dolphins (Tursiops truncatus).
Development and testing of species-specific ELISA assays to measure IFN-γ and TNF-α in bottlenose dolphins (Tursiops truncatus).
PloS one (2018-01-06)
Kirsten C Eberle, Stephanie K Venn-Watson, Eric D Jensen, Joanna LaBresh, Yvonne Sullivan, Laura Kakach, Randy E Sacco
ABSTRACT
Monitoring the immune status of cetaceans is important for a variety of health conditions. Assays to quantify cytokines, especially pro-inflammatory cytokines, could be employed, in addition to currently available diagnostic assays, to screen for alterations in the health status of an animal. Though a number of immunological assays are readily available for humans and mice, specific assays for many veterinary species, including cetaceans such as bottlenose dolphins (Tursiops truncatus), are more limited. Herein, we describe the development of IFN-gamma (IFN-γ) and TNF-alpha (TNF-α) enzyme-linked immunosorbent assays (ELISAs) specific to bottlenose dolphins. Utilizing these assays, we monitored the immune status of bottlenose dolphins from a managed population over a period of eleven months. The ELISA assays developed for bottlenose dolphins were used to measure IFN-γ and TNF-α in serum or in culture supernatants from peripheral blood mononuclear cells (PBMCs) stimulated with varying concentrations of mitogens concanavalin A (ConA) or phytohemagglutinin (PHA). Induction of TNF-α in PBMC cultures was consistently highest with 1 μg/mL ConA, while 1 μg/mL PHA induced the highest secretion of IFN-γ. Serum levels of TNF-α and IFN-γ remained relatively constant for each animal over the time period examined. CBC and plasma chemistry variables measured concurrently in the bottlenose dolphins were then examined as independent predictors of cytokine levels. We found these clinical variables were more likely to predict linear changes in serum IFN-γ and TNF-α levels compared to concentrations of these cytokines in mitogen-stimulated PBMC culture supernatants. Cytokine assays developed will be of substantial benefit in monitoring bottlenose dolphin health as an adjunct to currently available diagnostic tests.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sigma-Aldrich
Carbonate-Bicarbonate Buffer, capsule
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Phytohemagglutinin PHA-P, lyophilized powder (contains ~50% buffer salts as sodium phosphate and NaCl), BioReagent, suitable for cell culture