MilliporeSigma
  • Evolution of technology for molecular genotyping in blood group systems.
Evolution of technology for molecular genotyping in blood group systems.
The Indian journal of medical research (2018-01-23)
Ajit Gorakshakar, Harita Gogri, Kanjaksha Ghosh
ABSTRACT
The molecular basis of the blood group antigens was identified first in the 1980s and 1990s. Since then the importance of molecular biology in transfusion medicine has been described extensively by several investigators. Molecular genotyping of blood group antigens is one of the important aspects and is successfully making its way into transfusion medicine. Low-, medium- and high-throughput techniques have been developed for this purpose. Depending on the requirement of the centre like screening for high- or low-prevalence antigens where antisera are not available, correct typing of multiple transfused patients, screening for antigen-negative donor units to reduce the rate of alloimmunization, etc. a suitable technique can be selected. The present review discusses the evolution of different techniques to detect molecular genotypes of blood group systems and how these approaches can be used in transfusion medicine where haemagglutination is of limited value. Currently, this technology is being used in only a few blood banks in India. Hence, there is a need for understanding this technology with all its variations.