MilliporeSigma
  • Variability of intended copies for etanercept (Enbrel®): Data on multiple batches of seven products.
Variability of intended copies for etanercept (Enbrel®): Data on multiple batches of seven products.
mAbs (2017-10-12)
Brian Hassett, Morton Scheinberg, Gilberto Castañeda-Hernández, Mengtao Li, Uppuluri R K Rao, Ena Singh, Ehab Mahgoub, Javier Coindreau, Julie O'Brien, Steven M Vicik, Brian Fitzpatrick
ABSTRACT
Fusion protein and monoclonal antibody-based tumor necrosis factor (TNF) inhibitors represent established treatment options for a range of inflammatory diseases. Regulatory authorities have outlined the structural characterization and clinical assessments necessary to establish biosimilarity of a new biotherapeutic product with the innovator biologic drug. Biologic products that would not meet the minimum World Health Organization's standard for evaluation of similar biotherapeutic products are available in some countries; in some cases relevant data to assess biosimilarity and appropriate regulatory approval pathways are lacking. Batches of seven intended copy (IC) products for etanercept (Enbrel®) were subjected to a subset of test methods used in the routine release and heightened characterization of Enbrel®, to determine key attributes of identity, quality, purity, strength, and activity. While a number of quality attributes of the IC lots tested met the release specifications for Enbrel®, none fell within these limits across all methods performed, and there were no IC lots that satisfied the criteria typically applied by the innovator to support comparability with Enbrel®. Although the consequences of these differences are largely unknown, the potential for unanticipated clinical outcomes should not be overlooked.
Sigma-Aldrich
TWEEN® 80, BioXtra, viscous liquid