MilliporeSigma
  • Effect of negative dietary cation-anion differences on carcass characteristics and beef tenderness of Japanese Black steers.
Effect of negative dietary cation-anion differences on carcass characteristics and beef tenderness of Japanese Black steers.
Animal science journal = Nihon chikusan Gakkaiho (2017-09-16)
Kyohei Ishida, Kazuo Hirano
ABSTRACT
Lowering dietary cation-anion differences (DCAD) can enhance responsiveness to Ca-homeostatic hormones and increase Ca availability, which might have potential to activate a Ca-dependent protease, calpain, and to enhance postmortem myofibrillar proteolysis. In this study, we investigated the effects of DCAD manipulation on calpain activity and beef tenderness in Japanese Black cattle which are characterized by their high marbling. Thirty-six Japanese Black steers were allotted to one of two treatments: (i) control (CON; DCAD +6.09 mEq/100 g of dry matter (DM)) or (ii) negative DCAD (NEGD; DCAD -8.27 mEq/100 g DM) for 70 days before slaughter. Lowering DCAD decreased DM and energy intake (P < 0.01) even though it did not negatively affect the growth performance or carcass characteristics. In NEGD, urine pH was decreased by acidification caused by the negative DCAD (P < 0.01). Calpain activities tended to be improved in NEGD (P = 0.09), but Warner-Bratzler shear force values were not affected by treatment. Although calpain activities tended to improve, lowering DCAD to -8.27 for 70 days before slaughter was insufficient to enhance beef tenderness in Japanese Black steers.
Sigma-Aldrich
Di-(4-chlorobenzyl)azodicarboxylate, 97%