MilliporeSigma
N and O isotope (δ
Rapid communications in mass spectrometry : RCM (2017-11-14)
Leonard I Wassenaar, Cedric Douence, Mark A Altabet, Pradeep K Aggarwal
ABSTRACT
The nitrogen and oxygen (δ δ NO The Cd-azide method yielded excellent results for routine determination of δ
Sigma-Aldrich
Wheaton glass serum bottle, volume 125 mL, clear, O.D. 20 mm, diam. × H 54 mm × 107 mm