MilliporeSigma
Self-Assembled Nanomicelles as MRI Blood-Pool Contrast Agent.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-11-10)
Andrej Babič, Vassily Vorobiev, Céline Xayaphoummine, Gaëlle Lapicorey, Anne-Sophie Chauvin, Lothar Helm, Eric Allémann
ABSTRACT
Gadolinium-loaded nanomicelles show promise as future magnetic resonance imaging (MRI) contrast agents (CAs). Their increased size and high gadolinium (Gd) loading gives them an edge in proton relaxivity over smaller molecular Gd-complexes. Their size and stealth properties are fundamental for their long blood residence time, opening the possibility for use as blood-pool contrast agents. Using l-tyrosine as a three-functional scaffold we synthesized a nanostructure building block 8. The double C18 aliphatic chain on one side, Gd-1,4,7,10-tetraazacyclododecane-1-4-7-triacetic acid (Gd-DO3A) with access to bulk water in the center and 2 kDa PEG on the hydrophilic side gave the amphiphilic properties required for the core-shell nanomicellar architecture. The self-assembly into Gd-loaded monodispersed 10-20 nm nanomicelles occurred spontaneously in water. These nanomicelles (Tyr-MRI) display very high relaxivity at 29 mm
Sigma-Aldrich
4-(Chloromethyl)benzoyl chloride, 97%
Sigma-Aldrich
Dioctadecylamine, ≥99.0% (NT)