MilliporeSigma
  • Biodegradation of fluoranthene by Paenibacillus sp. strain PRNK-6: a pathway for complete mineralization.
Biodegradation of fluoranthene by Paenibacillus sp. strain PRNK-6: a pathway for complete mineralization.
Archives of microbiology (2017-09-25)
Pooja V Reddy, T B Karegoudar, T R Monisha, I Mukram, Anand S Nayak
ABSTRACT
A high-efficiency fluoranthene-degrading bacterium Paenibacillus sp. PRNK-6 was isolated from PAH-contaminated soil. The strain degrades 96% (240 mg l
Sigma-Aldrich
2-Carboxybenzaldehyde, 97%
Sigma-Aldrich
9-Hydroxyfluorene, 96%