MilliporeSigma
  • Decrease in male mouse fertility by hydrogen sulfide and/or ammonia can Be inheritable.
Decrease in male mouse fertility by hydrogen sulfide and/or ammonia can Be inheritable.
Chemosphere (2017-12-05)
Weidong Zhang, Yong Zhao, Pengfei Zhang, Yanan Hao, Shuai Yu, Lingjiang Min, Lan Li, Dongxue Ma, Liang Chen, Bao Yi, Xiangfang Tang, Qingshi Meng, Lei Liu, Shukun Wang, Wei Shen, Hongfu Zhang
ABSTRACT
Numerous epidemiological studies suggest that air pollutants cause a decline in the quality of human spermatozoa and thus a reduction in fertility. However, the exact cause of infertility remains unknown. Air pollution gases, such as NH
Sigma-Aldrich
Human LHB / Lutropin Subunit Beta ELISA Kit