MilliporeSigma
  • Rapid Mobilization Reveals a Highly Engraftable Hematopoietic Stem Cell.
Rapid Mobilization Reveals a Highly Engraftable Hematopoietic Stem Cell.
Cell (2017-12-12)
Jonathan Hoggatt, Pratibha Singh, Tiffany A Tate, Bin-Kuan Chou, Shruti R Datari, Seiji Fukuda, Liqiong Liu, Peter V Kharchenko, Amir Schajnovitz, Ninib Baryawno, Francois E Mercier, Joseph Boyer, Jason Gardner, Dwight M Morrow, David T Scadden, Louis M Pelus
ABSTRACT
Hematopoietic stem cell transplantation is a potential curative therapy for malignant and nonmalignant diseases. Improving the efficiency of stem cell collection and the quality of the cells acquired can broaden the donor pool and improve patient outcomes. We developed a rapid stem cell mobilization regimen utilizing a unique CXCR2 agonist, GROβ, and the CXCR4 antagonist AMD3100. A single injection of both agents resulted in stem cell mobilization peaking within 15 min that was equivalent in magnitude to a standard multi-day regimen of granulocyte colony-stimulating factor (G-CSF). Mechanistic studies determined that rapid mobilization results from synergistic signaling on neutrophils, resulting in enhanced MMP-9 release, and unexpectedly revealed genetic polymorphisms in MMP-9 that alter activity. This mobilization regimen results in preferential trafficking of stem cells that demonstrate a higher engraftment efficiency than those mobilized by G-CSF. Our studies suggest a potential new strategy for the rapid collection of an improved hematopoietic graft.
Sigma-Aldrich
Pertussis toxin from Bordetella pertussis, buffered aqueous glycerol solution