MilliporeSigma
  • Binding interactions of halogenated bisphenol A with mouse PPARα: In vitro investigation and molecular dynamics simulation.
Binding interactions of halogenated bisphenol A with mouse PPARα: In vitro investigation and molecular dynamics simulation.
Toxicology letters (2017-11-13)
Jie Zhang, Tiezhu Li, Tuoyi Wang, Tianzhu Guan, Hansong Yu, Zhuolin Li, Yongzhi Wang, Yongjun Wang, Tiehua Zhang
ABSTRACT
The binding of bisphenol A (BPA) and its halogenated derivatives (halogenated BPAs) to mouse peroxisome proliferator-activated receptor α ligand binding domain (mPPARα-LBD) was examined by a combination of in vitro investigation and in silico simulation. Fluorescence polarization (FP) assay showed that halogenated BPAs could bind to mPPARα-LBD* as the affinity ligands. The calculated electrostatic potential (ESP) illustrated the different charge distributions of halogenated BPAs with altered halogenation patterns. As electron-attracting substituents, halogens decrease the positive electrostatic potential and thereby have a significant influence on the electrostatic interactions of halogenated BPAs with mPPARα-LBD*. The docking results elucidated that hydrophobic and hydrogen-bonding interactions may also contribute to stabilize the binding of the halogenated BPAs to their receptor molecule. Comparison of the calculated binding energies with the experimentally determined affinities yielded a good correlation (R
Sigma-Aldrich
4,4′-(Hexafluoroisopropylidene)diphenol, 97%