MilliporeSigma
  • Enhanced fluidity liquid chromatography of inulin fructans using ternary solvent strength and selectivity gradients.
Enhanced fluidity liquid chromatography of inulin fructans using ternary solvent strength and selectivity gradients.
Analytica chimica acta (2017-12-20)
Raffeal Bennett, Susan V Olesik
ABSTRACT
The value of exploring selectivity and solvent strength ternary gradients in enhanced fluidity liquid chromatography (EFLC) is demonstrated for the separation of inulin-type fructans from chicory. Commercial binary pump systems for supercritical fluid chromatography only allow for the implementation of ternary solvent strength gradients which can be restrictive for the separation of polar polymeric analytes. In this work, a custom system was designed to extend the capability of EFLC to allow tuning of selectivity or solvent strength in ternary gradients. Gradient profiles were evaluated using the Berridge function (RF