MilliporeSigma
  • LINE-1 couples EMT programming with acquisition of oncogenic phenotypes in human bronchial epithelial cells.
LINE-1 couples EMT programming with acquisition of oncogenic phenotypes in human bronchial epithelial cells.
Oncotarget (2017-12-22)
Elsa M Reyes-Reyes, Ivan Aispuro, Marco A Tavera-Garcia, Matthew Field, Sara Moore, Irma Ramos, Kenneth S Ramos
ABSTRACT
Although several lines of evidence have established the central role of epithelial-to-mesenchymal-transition (EMT) in malignant progression of non-small cell lung cancers (NSCLCs), the molecular events connecting EMT to malignancy remain poorly understood. This study presents evidence that Long Interspersed Nuclear Element-1 (LINE-1) retrotransposon couples EMT programming with malignancy in human bronchial epithelial cells (BEAS-2B). This conclusion is supported by studies showing that: 1) activation of EMT programming by TGF-β1 increases LINE-1 mRNAs and protein; 2) the lung carcinogen benzo(a)pyrene coregulates TGF-β1 and LINE-1 mRNAs, with LINE-1 positioned downstream of TGF-β1 signaling; and, 3) forced expression of LINE-1 in BEAS-2B cells recapitulates EMT programming and induces malignant phenotypes and tumorigenesis
Sigma-Aldrich
MISSION® esiRNA, targeting human SNAI1