MilliporeSigma
  • RUNX2 promotes epithelial differentiation of ADSCs and burn wound healing via targeting E-cadherin.
RUNX2 promotes epithelial differentiation of ADSCs and burn wound healing via targeting E-cadherin.
Oncotarget (2018-02-09)
Qiang Li, Han Zhao, Sizhan Xia, Hanxiao Wei, Feifei Chen, Peisheng Jin
ABSTRACT
Epithelial differentiation of adipose-derived stem cells (ADSCs) is mediated by sophisticated interactions of various molecular functions and biological processes, including transcriptional regulation. Runt-related transcription factor 2 (RUNX2) increases osteoblast and adipocyte differentiation of ADSCs. However, the role of RUNX2 in epithelial differentiation of ADSCs is unknown. We first showed that ADSCs possess the potential to differentiate into epithelial lineage. Then, we employed the effect of RUNX2 on epithelial differentiation of ADSCs. Our data showed that RUNX2 promoted epithelial differentiation of ADSCs. Overexpression or knockdown of RUNX2 resulted in increase or decrease of E-cadherin expression, respectively. Abatement of E-cadherin in ADSCs attenuated RUNX2-activated epithelial conversion of ADSCs and epithelial markers cytokeratin 18 (CK18) and zonula occludens protein-1 (ZO-1). We also evaluated the effect of RUNX2 on burn wound healing
Sigma-Aldrich
MISSION® esiRNA, targeting human RUNX2