MilliporeSigma
  • New approach for vertical bone regeneration using in situ gelling and sustained BMP-2 releasing poly(phosphazene) hydrogel system on peri-implant site with critical defect in a canine model.
New approach for vertical bone regeneration using in situ gelling and sustained BMP-2 releasing poly(phosphazene) hydrogel system on peri-implant site with critical defect in a canine model.
Journal of biomedical materials research. Part B, Applied biomaterials (2017-03-24)
Bo-Bae Seo, Hae-Im Chang, Hyuck Choi, Jeong-Tae Koh, Kwi-Dug Yun, Jae-Yeol Lee, Soo-Chang Song
ABSTRACT
An injectable hydrogel system with sustained bone morphogenetic protein 2 (BMP-2) release ability was developed for vertical bone regeneration at peri-implant sites and enhanced osseointegration of dental implants. In three young male beagle dogs, a pair of defects was created on both sides of the mandibular bone. Next, two implants were transplanted into each defect. In situ gelling polymer solutions with or without BMP-2 were applied to cover the implants and mandibular defects. The effects of the in situ gelling and sustained BMP-2 releasing (IGSR) hydrogel system on peri-implant bone regeneration were evaluated by radiologic examination, micro-computed tomography, and histomorphometric analysis. Twelve weeks after the treatment, significant bone generation at the peri-implant site occurred following BMP-2/IGSR hydrogel treatment. Bone volume and mineral density were increased by 1.7- and 1.3-fold, respectively (p < 0.01 and 0.05 vs. control, respectively) for the BMP-2/IGSR hydrogel system. And, 0.57-0.31 mm vertical bone generation was observed at the peri-implant site for the BMP-2/IGSR hydrogel system, while rare vertical bone generation occurred in the control group. The BMP-2/IGSR hydrogel system significantly increased bone to implant contact % between induced bone and existing bone (p < 0.05 and 0.01 vs. control). These vertical bone regeneration and higher osseointegration levels demonstrated the effectiveness of the BMP-2/IGSR hydrogel system. © 2017 Wiley Periodicals, Inc. J Biomed Mater Res Part B: Appl Biomater, 106B: 751-759, 2018.
Sigma-Aldrich
Cefazolin sodium salt, 89.1-110.1%
Pricing and availability is not currently available.