MilliporeSigma
  • Topically Applied Carvedilol Attenuates Solar Ultraviolet Radiation Induced Skin Carcinogenesis.
Topically Applied Carvedilol Attenuates Solar Ultraviolet Radiation Induced Skin Carcinogenesis.
Cancer prevention research (Philadelphia, Pa.) (2017-09-16)
Kevin M Huang, Sherry Liang, Steven Yeung, Etuajie Oiyemhonlan, Kristan H Cleveland, Cyrus Parsa, Robert Orlando, Frank L Meyskens, Bradley T Andresen, Ying Huang
ABSTRACT
In previous studies, the β-blocker carvedilol inhibited EGF-induced epidermal cell transformation and chemical carcinogen-induced mouse skin hyperplasia. As exposure to ultraviolet (UV) radiation leads to skin cancer, the present study examined whether carvedilol can prevent UV-induced carcinogenesis. Carvedilol absorbs UV like a sunscreen; thus, to separate pharmacological from sunscreen effects, 4-hydroxycarbazole (4-OHC), which absorbs UV to the same degree as carvedilol, served as control. JB6 P
Sigma-Aldrich
4-Hydroxycarbazole, 95%