MilliporeSigma
  • Influence of the Chemical Structure on Odor Qualities and Odor Thresholds of Halogenated Guaiacol-Derived Odorants.
Influence of the Chemical Structure on Odor Qualities and Odor Thresholds of Halogenated Guaiacol-Derived Odorants.
Frontiers in chemistry (2018-01-13)
Florian Juhlke, Katja Lorber, Maria Wagenstaller, Andrea Buettner
ABSTRACT
Chlorinated guaiacol derivatives are found in waste water of pulp mills using chlorine in the bleaching process of wood pulp. They can also be detected in fish tissue, possibly causing off-odors. To date, there is no systematic investigation on the odor properties of halogenated guaiacol derivatives. To close this gap, odor thresholds in air and odor qualities of 14 compounds were determined by gas chromatography-olfactometry. Overall, the investigated compounds elicited smells that are characteristic for guaiacol, namely
Sigma-Aldrich
2-Iodophenol, 98%
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
4-Iodo-2-methoxyphenol, 97%