MilliporeSigma
  • Placental growth factor signaling regulates isoform splicing of vascular endothelial growth factor A in the control of lung cancer cell metastasis.
Placental growth factor signaling regulates isoform splicing of vascular endothelial growth factor A in the control of lung cancer cell metastasis.
Molecular and cellular biochemistry (2017-09-02)
Zanfeng Wang, Tingwei Liu
ABSTRACT
Vascular endothelial growth factor (VEGF) family members play critical and complex roles in the regulation of cancer vascularization and metastasis. The exact molecular control of lung cancer metastasis by VEGF family members is not completely understood. Here, we showed that specimens from non-small cell lung cancer (NSCLC) contained significantly higher levels of placental growth factor (PlGF) than paired non-cancer tissue (p < 0.05, N = 25). Moreover, higher levels of PlGF were detected in NSCLC specimens from the patients who had distal metastases than those who had not. High-PlGF levels appeared to be associated with poor patient survival. In vitro, PlGF dose-dependently increased the ratio of pro-angiogenic VEGF isoform (VEGF