MilliporeSigma
  • Mir-144-3p Promotes Cell Proliferation, Metastasis, Sunitinib Resistance in Clear Cell Renal Cell Carcinoma by Downregulating ARID1A.
Mir-144-3p Promotes Cell Proliferation, Metastasis, Sunitinib Resistance in Clear Cell Renal Cell Carcinoma by Downregulating ARID1A.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-10-27)
Wen Xiao, Ning Lou, Hailong Ruan, Lin Bao, Zhiyong Xiong, Changfei Yuan, Junwei Tong, Guanghua Xu, Yali Zhou, Yan Qu, Wenjun Hu, Yaoying Gao, Zeyuan Ru, Lei Liu, Haibing Xiao, Ke Chen, Hongmei Yang, Xiaoping Zhang
ABSTRACT
We previously performed microRNA (miRNA) microarray to identify effective indicators of clear cell renal cell carcinoma (ccRCC) tissue samples and preoperative/postoperative plasma in which we identified miR-144-3p as an oncomiRNA. However, the molecular mechanism of miR-144-3p remains unclear. This study aims to explore the roles of miR-144-3p in the invasion, migration and Sunitinib-resistance in ccRCC and to elucidate the underlying mechanisms. Gain and loss of function approaches were used to investigate the cell proliferation, cycle distribution, clonogenicity, migration, invasion, chemosensitivity of miR-144-3p in vitro. The xenograft model was used to assess the effects of miR-144-3p overexpression on tumorigenesis. Bioinformatics analysis and dual-luciferase reporter assay were used to indentify AT-rich interactive domain 1A (ARID1A) as a direct target gene of miR-144-3p. Quantitative RT-PCR, Western blotting, and immunohistochemical (IHC) staining were used to explore ARID1A expression level of the mRNA and protein. We found that miR-144-3p overexpression enhanced cell proliferation, clonogenicity, migration, invasion, and chemoresistance in ccRCC cells. Notably, the oncotumor activities of miR-144-3p were mediated by repressing the expression of ARID1A. The downregulation of ARIDIA could promote the function of miR-144-3p in cell proliferation, metastasis and chemoresistance. Consistently, ARID1A mRNA and protein levels were decreased in ccRCC and in nude mice, and they negatively correlated with miR-144-3p. Higher miR-144-3p may enhance malignancy and resistance to Sunitinib in ccRCC by targeting ARID1A, the observations may uncover novel strategies of ccRCC treatment.