- Potential of rare actinomycetes in the production of metabolites against multiple oxidant agents.
Potential of rare actinomycetes in the production of metabolites against multiple oxidant agents.
Actinobacteria are a precious source of novel bioactive metabolites with potential pharmaceutical applications. Representatives of 11 genera of rare Actinobacteria were selected for the evaluation of antioxidant activity. Fermentation broths of the Actinobacteria were extracted and dosage of 10 to 2000āµg/mL were applied for in vitro antioxidant-related bioassays. Cytotoxicity was assessed at the concentration of 2.5-20āµg/mL. In the DPPH scavenging activity, 15 out of 52 extracts showed 17.0-26.8% activity in quantitative evaluation. Metabolites of five prominent antioxidant producing strains protected the DNA (pUC19) against UV-induced photolyzed H The presence of nonidentical metabolites might be responsible for antioxidant and enzyme inhibitory activities of S. ecbatanensis, newly described actinobacterium in family Pseudonocardiaceae. The scavenging of the free electrons, protection of DNA and model yeast cells against oxidative stress, in addition to the inhibition of the oxidating enzymes are the main mechanisms of the antioxidant effect of the introduced resource in this study.