MilliporeSigma
C-terminal cleavage of the LH1 α-polypeptide in the Sr
Photosynthesis research (2017-05-12)
Yukihiro Kimura, Tomoaki Kawakami, Teruhisa Arikawa, Yong Li, Long-Jiang Yu, Takashi Ohno, Michael T Madigan, Zheng-Yu Wang-Otomo
ABSTRACT
The light-harvesting 1 reaction center (LH1-RC) complex in the thermophilic purple sulfur bacterium Thermochromatium (Tch.) tepidum binds Ca ions as cofactors, and Ca-binding is largely involved in its characteristic Q