MilliporeSigma
  • Palladium-catalyzed carbonylation of yne esters leading to gamma-alkylidene alpha,beta-unsaturated gamma-lactones.
Palladium-catalyzed carbonylation of yne esters leading to gamma-alkylidene alpha,beta-unsaturated gamma-lactones.
Organic letters (2005-09-24)
Yasuyuki Harada, Yoshiya Fukumoto, Naoto Chatani
ABSTRACT
[reaction: see text] The reaction of yne esters with carbon monoxide (1 atm) in the presence of palladium complexes gives bicyclic unsaturated lactone derivatives in good to high yields. The 2-pyridinyloxy group is a good leaving group among leaving groups examined.
Sigma-Aldrich
Allylpalladium(II) chloride dimer, 98%